missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human INSRR Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | INSRR/IR-related receptor |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
INSRR/IR-related receptor Polyclonal specifically detects INSRR/IR-related receptor in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
INSRR/IR-related receptor | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC 2.7.10, EC 2.7.10.1, insulin receptor-related protein, insulin receptor-related receptor, IRR, IRRIR-related receptor | |
INSRR | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P14616 | |
3645 | |
Mouse myeloma cell line NS0-derived recombinant human INSRR Leu24-Leu923 Accession # P14616 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human INSRR in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human INSR is observed. |
For Research Use Only