missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InPro™ 3253i/SG pH Electrodes

Brand:  Mettler Toledo™ 52005379

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12737681

  • POA / Each

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

12mm
Solution Ground
0 to 14
InPro® 3253i/SG pH Electrode
CE Certified
325mm
Glass
0 to 100°C
K8S
Videos