missing translation for 'onlineSavingsMsg'
Learn More

Inosine, 99%, Thermo Scientific Chemicals

Catalog Number 10553884
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
58-63-9
C10H12N4O5
268.23
MFCD00066770
UGQMRVRMYYASKQ-YPLCUDRINA-N
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
6021
CHEBI:17596
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
58-63-9
C10H12N4O5
268.23
MFCD00066770
UGQMRVRMYYASKQ-YPLCUDRINA-N
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
6021
CHEBI:17596
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
Melting Point 212°C to 213°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 g
Beilstein 31, 25
Merck Index 15, 5018
Specific Rotation Condition − 49.20 (18.00°C c=1,H2O)
Specific Rotation -49.2
Solubility Information Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol
Spectrophotometry 98 to 102% (lambda max. 248.5nm,pH 7) Molar absorptivity 12200+/-2%,on dry substance
Transmittance 98% min. (c=1, H2O, 430nm, 1cm cell)
Formula Weight 268.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Inosine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'