missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Initial Calibration Blank/Continuing Calibration Blank(ICB/CCB), Specpure™

Brand:  Alfa Aesar™ 035259.AP

Code : NEW

Additional Details : Weight : 0.00050kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Product Code. 15433575

  • £57.80 / 500mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Specifications

Initial Calibration Blank/Continuing Calibration Blank(ICB/CCB)
Liquid
MFCD00147250
500mL
Specpure™
Matrix: 5% HCl/1% HNO3
UN3264
Videos
SDS