missing translation for 'onlineSavingsMsg'
Learn More

INH1, Tocris Bioscience™

Hec1 inhibitor; causes arrest of mitosis

Brand:  Tocris (Bio-Techne) 355110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433184

  • £138.18 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Hec1 inhibitor. Binds Hec1, inhibiting its association with Nek2 and kinetochores. Causes arrest of mitosis and inhibits proliferation in breast cancer cell lines (GI50= 10 - 20 μM).
Specifications

Chemical Identifiers

313553-47-8
308.399
n-4-2,4-dimethylphenyl thiazol-2-yl benzamide hydrochloride, c18h16n2os.hcl, n-4-2,4-dimethylphenyl-2-thiazolyl benzamide hydrochloride, n-4-2,4-dimethylphenyl-2-thiazolyl benzamidehydrochloride, n-4-2,4-dimethylphenyl-1,3-thiazol-2-yl benzamide;hydrochloride, n-4-2,4-dimethylphenyl-1,3-thiazol-2-yl benzamide-hydrogen chloride 1/1
N-[4-(2,4-dimethylphenyl)-1,3-thiazol-2-yl]benzamide
C18H16N2OS
JPMOKRWIYQGMJL-UHFFFAOYSA-N
959043
CC1=CC(=C(C=C1)C2=CSC(=N2)NC(=O)C3=CC=CC=C3)C

Specifications

INH1
313553-47-8
n-4-2,4-dimethylphenyl thiazol-2-yl benzamide hydrochloride, c18h16n2os.hcl, n-4-2,4-dimethylphenyl-2-thiazolyl benzamide hydrochloride, n-4-2,4-dimethylphenyl-2-thiazolyl benzamidehydrochloride, n-4-2,4-dimethylphenyl-1,3-thiazol-2-yl benzamide;hydrochloride, n-4-2,4-dimethylphenyl-1,3-thiazol-2-yl benzamide-hydrogen chloride 1/1
CC1=CC(=C(C=C1)C2=CSC(=N2)NC(=O)C3=CC=CC=C3)C
308.399
344.86
10mg
C18H16N2OS
JPMOKRWIYQGMJL-UHFFFAOYSA-N
N-[4-(2,4-dimethylphenyl)-1,3-thiazol-2-yl]benzamide
959043
>99%
Videos
Documents