missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Sorensen's Buffer (concentrated)
Sorensen’s Buffer (concentrated)
Brand: Novacyt - Microgen Bioproducts M1
Additional Details : Weight : 0.50000kg
Description
- Sorensen's Buffer (concentrated)
Specifications
25 mL Sorensens Buffer | |
Clear | |
Liquid | |
Bottle |
Approx. 100°C | |
6.8 | |
Sorensens Buffer |