missing translation for 'onlineSavingsMsg'
Learn More

Manutan Industrial locker with plinth, three nest, one tier, cylinder, grey, 1850mm x 930mm x 500mm

Brand:  Manutan 1062O054

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12825246

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Smart range of lockers with excellent ventilation, ideal for workwear storage. Prevent unauthorized access with concealed hinges and a choice of either a cylinder or hasp lock. Vent holes at the top and bottom of doors. Fitted with 150mm plinth. One tier lockers with hat shelf, hanging rail and three coat hooks. Two tier lockers with hanging rail and three coat hooks. Label holder on each door with powder coated finish.
  • Three lockers supplied
  • One tier
  • Cylinder lock
  • Grey door
  • Dimensions (H x W x D): 1850mm x 930mm x 500mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers