missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-6-carboxaldehyde, 97%, ACROS Organics™
£65.90 - £191.00
Chemical Identifiers
CAS | 1196-70-9 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.16 |
MDL Number | MFCD02179596 |
InChI Key | VSPBWOAEHQDXRD-UHFFFAOYSA-N |
Synonym | indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole |
PubChem CID | 2773435 |
IUPAC Name | 1H-indole-6-carbaldehyde |
SMILES | C1=CC(=CC2=C1C=CN2)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10492950
|
Acros Organics
435512500 |
250mg | Glass bottle |
£65.90
250mg |
Estimated Shipment 08-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10018049
|
Acros Organics
435510010 |
1g | Glass bottle |
£191.00
1g |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
1196-70-9 | |
145.16 | |
VSPBWOAEHQDXRD-UHFFFAOYSA-N | |
2773435 | |
C1=CC(=CC2=C1C=CN2)C=O |
C9H7NO | |
MFCD02179596 | |
indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole | |
1H-indole-6-carbaldehyde |
Specifications
Indole-6-carboxaldehyde | |
Glass bottle | |
129°C to 131°C | |
1196-70-9 | |
C9H7NO | |
indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole | |
C1=CC(=CC2=C1C=CN2)C=O | |
145.16 | |
145.16 | |
97% |
Authentic | |
Brown to Yellow | |
250mg | |
96% min. (HPLC) | |
MFCD02179596 | |
VSPBWOAEHQDXRD-UHFFFAOYSA-N | |
1H-indole-6-carbaldehyde | |
2773435 | |
Crystalline Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Indole-6-carboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.