missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldoxime, 97%, Thermo Scientific™

Catalog Number p-7036459
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2592-05-4
MFCD00022737
This item is not returnable. View return policy
2592-05-4
MFCD00022737

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Molecular Formula C12H20O6
Quantity 250 mg
Synonym 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
InChI Key KEJGAYKWRDILTF-VVULQXIFSA-N
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C
IUPAC Name (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
Molecular Weight (g/mol) 260.29
PubChem CID 7157054
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Indole-3-carboxaldoxime
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'