missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10112964
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19005-93-7
C9H7NO
145.16
MFCD03001425
SBNOTUDDIXOFSN-UHFFFAOYSA-N
indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, indole-2-carbaldehyde, 2-formylindole, 2-formyl-1h-indole, 1h-indolecarboxaldehyde, unii-r5n9cd7dlm, r5n9cd7dlm, methanol, 2h-indol-2-ylidene-, 1e-9ci, indolaldehyd
96389
1H-indole-2-carbaldehyde
O=CC1=CC2=CC=CC=C2N1
This item is not returnable. View return policy
19005-93-7
C9H7NO
145.16
MFCD03001425
SBNOTUDDIXOFSN-UHFFFAOYSA-N
indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, indole-2-carbaldehyde, 2-formylindole, 2-formyl-1h-indole, 1h-indolecarboxaldehyde, unii-r5n9cd7dlm, r5n9cd7dlm, methanol, 2h-indol-2-ylidene-, 1e-9ci, indolaldehyd
96389
1H-indole-2-carbaldehyde
O=CC1=CC2=CC=CC=C2N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Color Gray to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 145.16
Percent Purity 97%
Physical Form Solution
Chemical Name or Material Indole-2-carboxaldehyde
compliance-icons
Product Identifier
  • Indole-2-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'