Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2 with certificate
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand:  Fisherbrand™
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15555159
  • £63.12 / 1 unit
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Specifications
F2
50 g
Cylindrical
Stainless Steel
50 g