missing translation for 'onlineSavingsMsg'
Learn More

Indium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

Brand:  Fisher Chemical S026100

Code : 04

Additional Details : Weight : 0.10000kg

Quantity 100mL

 View more versions of this product

Product Code. 12953424

  • £53.46 / 100mL


Estimated Shipment
05-04-2018
Add to basket

Specifications

Chemical Identifiers

1g/L
HNO3

Specifications

1g/L
HNO3
100mL
Videos