missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium powder, -100 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Color | Silver to Gray |
Melting Point | 156.6°C |
Boiling Point | 2072°C |
Density | 7.31 g/mL |
UN Number | UN3089 |
Quantity | 5 g |
Formula Weight | 114.82 |
Percent Purity | 99.99% |
Mesh Size | −100 Mesh |
Odor | Odorless |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'