missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9999% (metals basis)

Brand:  Alfa Aesar™ 010617.90

Code : NEW

Additional Details : CAS Number : 7440-74-6 Weight : 0.25000kg

Quantity 5 x 50g

 View more versions of this product

Product Code. 11330789

  • £1912.10 / 250g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Indium ingot
Puratronic™
MFCD00134048
7440-74-6
114.82

Specifications

Indium ingot
7440-74-6
Polycrystalline Ingot
(metals basis)
MFCD00134048
5 x 50g
Puratronic™
99.9999%
114.82
Odorless
Videos
SDS