missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051843
28.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
38218-24-5
C9H21InO3
292.082
MFCD00210318
OVZUSPADPSOQQN-UHFFFAOYSA-N
triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
57346963
indium(3+);propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
38218-24-5
C9H21InO3
292.082
MFCD00210318
OVZUSPADPSOQQN-UHFFFAOYSA-N
triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
57346963
indium(3+);propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]

Precursor used in the preparation of indium oxide containing films

Indium(III) isopropoxide acts as a precursor for the preparation of indium oxide containing films. It finds application in nano-materials and thin films display. Furthermore, it is used in imaging catalysis and in preparation of sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Melting Point 145°C (decomposition)
Color White
Linear Formula In[OCH(CH3)2]3
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohols,ketones and esters.
Formula Weight 292.08
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Indium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'