missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4824752
Click to view available options
Quantity:
10 g
50 g
250 g
10025-82-8
InCl3
MFCD00011058
10025-82-8
InCl3
MFCD00011058

Indium(III) chloride is used in organic reactions such as Friedel-crafts acylation and Diels-Alder reaction as a Lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Merck Index 14,4958
Solubility Information Soluble in water and tetrhydrofuran; Sparingly soluble in alcohol; Insoluble in ether.
Formula Weight 221.18
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.460
Assay (metals basis)
Chemical Name or Material Indium(III) chloride, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'