missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

Catalog Number p-7049286
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
14405-45-9
C15H21InO6
412.15
MFCD00013494
CMGVNFXMZKDHDG-UHFFFAOYSA-N
acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate
101644361
indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
This item is not returnable. View return policy
14405-45-9
C15H21InO6
412.15
MFCD00013494
CMGVNFXMZKDHDG-UHFFFAOYSA-N
acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate
101644361
indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Used in catalysts

Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 186°C (decomposition)
Quantity 1 g
Solubility Information Soluble in benzene; Insoluble in water.
Formula Weight 412.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indium(III) 2,4-pentanedionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'