missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(I) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Indium(I) chloride is used to prepare cage, dinuclear complexes (In-cyclodiphophazanes) which possess interesting metal-metal interactions, Semiconductor doping, phosphors-compound semiconductors, ITO thin-film, Basic raw material for compounding organic indium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 608°C |
Melting Point | ∼225°C |
Physical Form | Crystalline Lump |
Quantity | 5 g |
UN Number | UN3260 |
Solubility Information | Decomposes in water. |
Formula Weight | 150.27 |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Odor | Chlorine-like |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'