missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048502
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
13465-10-6
Cl3In
221.17
MFCD00148875
PSCMQHVBLHHWTO-UHFFFAOYSA-K
indium(3+) trichloride
[Cl-].[Cl-].[Cl-].[In+3]
This item is not returnable. View return policy
13465-10-6
Cl3In
221.17
MFCD00148875
PSCMQHVBLHHWTO-UHFFFAOYSA-K
indium(3+) trichloride
[Cl-].[Cl-].[Cl-].[In+3]

Indium(I) chloride is used to prepare cage, dinuclear complexes (In-cyclodiphophazanes) which possess interesting metal-metal interactions, Semiconductor doping, phosphors-compound semiconductors, ITO thin-film, Basic raw material for compounding organic indium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 608°C
Melting Point ∼225°C
Physical Form Crystalline Lump
Quantity 5 g
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 150.27
Percent Purity 99.999%
Grade Puratronic™
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 4.19 g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'