missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050749
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14280-53-6
MFCD00064630
indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci
This item is not returnable. View return policy
14280-53-6
MFCD00064630
indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 662°C
Melting Point 220°C
Physical Form Beads
Quantity 5 g
UN Number UN3260
Formula Weight 194.73
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Moisture Sensitive
Density 4.96g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'