missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium antimonide, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Indium antimonide finds use in infrared detectors, including FLIR systems, thermal imaging cameras, infrared astronomy and in infrared homing missile guidance systems, in fast transistors. It is used in thermal image detectors using photo-electromagnetic detectors or photodiodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 535°C |
Physical Form | Pieces |
Quantity | 2 g |
UN Number | UN1549 |
Merck Index | 14,4948 |
Solubility Information | Insoluble in water. |
Formula Weight | 236.57 |
Percent Purity | 99.999% |
Odor | Odorless |
Density | 5.75 g/mL at m.p. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'