Learn More
Tocris Bioscience™ Indiplon
Subtype-selective GABAA receptor positive allosteric modulator
Brand:  Tocris Bioscience™ 359710
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463264
  • £145.80 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Potent GABAA receptor positive allosteric modulator that acts at the benzodiazepine site (Ki values are 1.2 and 1.7 nM in rat frontal cortex and cerebellum respectively). Displays ∽ 10-fold selectivity for α1 subunit-containing receptors (EC50 values are 2.6, 24, 60 and 77 nM for α1β2γ2, α2β2γ2, α3β3γ2 and α5β2γ2 receptors respectively). Exhibits sedative, hypnotic, anxiolytic and anticonvulsant activity in vivo and is orally active.
Specifications
Specifications
Indiplon
>99
10mg
Indiplon
376.43