Learn More
Tocris Bioscience™ Indibulin
Microtubule destabilizer
Brand:  Tocris Bioscience™ 372850
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443504
  • £578.53 / 50mg
Estimated Shipment
28-06-2019
Description
Description
Microtubule destabilizer that blocks tubulin polymerization (IC50 = 0.3 μM). Blocks cell cycle progression at metaphase and displays antitumor activity against a variety of malignancies in vitro (IC50 values ranging from 0.036 - 0.285 μM). Induces complete tumor remission in the Yoshida AH13 rat sarcoma model in vivo. Lacks neurotoxicity due to an ability to discriminate between posttranslationally modified tubulin in mature neuronal microtubules and less-modified tubulin present in immature neuronal or nonneuronal microtubules. Displays oral availability and efficacy towards MDR tumor cells.
Specifications
Specifications
Indibulin
>98
50mg
Indibulin
389.83