missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indane, 95%, Thermo Scientific Chemicals
118.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 96%g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Molecular Formula | C9H10 |
Refractive Index | 1.5350 to 1.5380 (20°C, 589nm) |
Quantity | 500 g |
Beilstein | 05, IV, 1371 |
Merck Index | 15, 4971 |
Show More |
Safety and Handling
Product Identifier
- Indan
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'