missing translation for 'onlineSavingsMsg'
Learn More

Indane-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4228741
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
52205-85-3
C9H9ClO2S
216.679
SWLIXMXSCZYVTQ-UHFFFAOYSA-N
indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
3142583
2,3-dihydro-1H-indene-5-sulfonyl chloride
C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
52205-85-3
C9H9ClO2S
216.679
SWLIXMXSCZYVTQ-UHFFFAOYSA-N
indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
3142583
2,3-dihydro-1H-indene-5-sulfonyl chloride
C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 47.8°C to 48.8°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 216.69
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Indane-5-sulfonyl chloride
compliance-icons
Product Identifier
  • Indane-5-sulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'