Learn More
WTW™ Incubator Cabinets
Thermostat cabinets by WTW are excellent choices for respirometric measurements with OxiTop™ systems. In a specified temperature range, they yield a deviation of ±0,5 K in relation to the set temperature and can be adjusted in steps of 1° C. The heating with the radial blower for constant temperature is equipped with sockets for the stirring platforms.
Brand:  WTW™ 208381
Code : NEW
Additional Details : Weight : 30.00000kg
Product Code. 10671583
  • £1923.09 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
  • Incubator cabinet
  • For 2 BOD OxiTop Systems
For 2 BOD OxiTop Systems
Incubator Cabinet