Learn More
Alfa Aesar™ Inconel Cover for Crucible
Brand:  Alfa Aesar™ 036024.KT
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15451407
  • £24.38 / Each
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Specifications
45mL
For 35970 crucible
Inconel Crucible Cover
Inconel