missing translation for 'onlineSavingsMsg'
Learn More

Inconel™ 600 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 046745.FL

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15407305

  • £16.30 / Each
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications include heaters, stills, bubble towers and condensers for processing of fatty acids; evaporator tubes, tube sheets and flaking trays for the manufacture of sodium sulfide; and equipment for handling abietic acid in the manufacture of paper pulp. It is used for retorts, muffles, roller hearths and other furnace components and for heat-treating baskets and trays. The alloy is a standard material of construction for nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Inconel™ 600 gauze
Ni:Cr:Fe; 77:15.5:7.5 wt%
75 x 75 mm
40 Mesh
40 mesh woven from 0.25mm (0.01 in.) dia. wire
MFCD01741927
Insoluble in water.
Wire Cloth
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only