Learn More
Inconel|r 600 gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046791.FL
24.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
MFCD01741927 | |
200 Mesh |
200 mesh woven from 0.05mm (0.002 in.) dia. wire | |
75 x 75 mm | |
Wire Cloth |
RUO – Research Use Only