Learn More
Inconel|r 600 gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046791.GT
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
MFCD01741927 | |
200 Mesh |
200 mesh woven from 0.05mm (0.002 in.) dia. wire | |
150 x 150 mm | |
Wire Cloth |
Safety and Handling
- Inconel 600 gauze
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only