missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Inconel™ 600 gauze, 100 mesh woven from 0.102mm (0.004 in.) dia. wire

Brand:  Alfa Aesar™ 045069.FI

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50 x 50mm

 View more versions of this product

Product Code. 15436715

  • £18.20 / Each


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Inconel™ 600 gauze
100 mesh woven from 0.102mm (0.004 in.) dia. wire
MFCD01741927

Specifications

Ni:Cr:Fe; 77:15.5:7.5 wt%
100 Mesh
100 mesh woven from 0.102mm (0.004 in.) dia. wire
Inconel™ 600 gauze
50 x 50mm
MFCD01741927
Videos
SDS