Learn More
Inconel™ 600 gauze, 100 mesh woven from 0.102mm (0.004 in.) dia. wire, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045069.GH
17.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
Ni:Cr:Fe; 77:15.5:7.5 wt% | |
100 x 100 mm |
100 mesh woven from 0.102mm (0.004 in.) dia. wire | |
MFCD01741927 | |
100 Mesh |
RUO – Research Use Only