Learn More
Inconel|r 600 gauze, 10 mesh woven from 0.64mm (0.025in) dia wire, Thermo Scientific Chemicals
£17.20 - £102.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457215
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046453.FL
75 x 75 mm
£17.20
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
15467215
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046453.GT
150 x 150 mm
£29.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
15477215
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
046453.HB
300 x 300 mm
£102.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
Inconel is typically used in applications where high temperatures are present. Inconel superalloys also have excellent mechanical properties and are frequently used in the chemical and aerospace industries. It is common in gas turbine blades, seals, and as well as turbocharger rotors and seals, electric submersible well pump motor shafts, high temperature fasteners, chemical processing and pressure vessels, heat exchanger tubing, steam generators and core components in nuclear pressurized water reactors.