Learn More
Inconel|r 600 gauze, 10 mesh woven from 0.64mm (0.025in) dia wire, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046453.GT
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Inconel is typically used in applications where high temperatures are present. Inconel superalloys also have excellent mechanical properties and are frequently used in the chemical and aerospace industries. It is common in gas turbine blades, seals, and as well as turbocharger rotors and seals, electric submersible well pump motor shafts, high temperature fasteners, chemical processing and pressure vessels, heat exchanger tubing, steam generators and core components in nuclear pressurized water reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
Ni:Cr:Fe; 77:15.5:7.5 wt% | |
150 x 150 mm | |
10 Mesh |
10 mesh woven from 0.64mm (0.025 in.) dia. wire | |
MFCD01741927 | |
Insoluble in water. | |
Wire Cloth |
RUO – Research Use Only