Learn More
IMPG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564926
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LQAHHDRSERE SPFSGSSRQP DSLSSIENAV KYNPVYESHR AGCEKYEGPY PQHPFYSSAS GDVIGGLSRE EIRQMYESSE LSREEIQERM RVLELYANDP EFAAFVR Highest antigen sequence identity to the following orthologs - mouse 80%, rat 78%.
IMPG2 Polyclonal antibody specifically detects IMPG2 in Human samples. It is validated for Immunohistochemistry.Specifications
IMPG2 | |
Polyclonal | |
Unconjugated | |
IMPG2 | |
IMPG2; Interphotoreceptor matrix proteoglycan 2; interphotoreceptor matrix proteoglycan IPM 200; interphotoreceptor matrix proteoglycan of 200 kDa; IPM 200; IPM200; PG10.2; RP56; Rsbp; Sialoprotein associated with cones and rods proteoglycan; spacrcan; VMD5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZV3 | |
IMPG2 | |
Recombinant Human IMPG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only