missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Brand:  Thermo Scientific™ 469957

Code : 76

Additional Details : Weight : 2.28600kg

 View more versions of this product

Product Code. 10748091

  • £278.00 / Pack of 640
In Stock
to see stock.
Add to basket

Description

Description

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Specifications

Specifications

Hydrophobic
Strip Plate
1 x 8
Clear
Polystyrene
350 μL
Round, Flat Bottom (F-well)
160
640/Cs.
F8
No
Colorimetric
640 Cases
Microplate Strip
8
PolySorp
640
None
Lipids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers