Learn More
Immethridine dihydrobromide, Tocris Bioscience™
Potent H3 agonist, highly selective over H4
Brand:  Tocris (Bio-Techne) 231510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13441014
  • £124.20 / 10mg
Estimated Shipment
27-06-2019
Description
Description
Potent and highly selective histamine H3 receptor agonist (pEC50 = 9.74) that displays 300-fold selectivity over the H4 receptor (pKi values are 9.07 and 6.61 respectively). Does not bind to H1 or H2 receptors at concentrations up to 10 μM. .
Specifications
Chemical Identifiers
699020-93-4
321.016
immethridine dihydrobromide, 4-1h-imidazol-4-ylmethyl pyridine dihydrobromide, 4-1h-imidazol-5-ylmethyl pyridine;dihydrobromide, 4-1h-imidazol-5-yl methyl pyridine-hydrogen bromide 1/2
4-(1H-imidazol-5-ylmethyl)pyridine;dihydrobromide
C9H11Br2N3
CYNKWHIKNDIVDR-UHFFFAOYSA-N
16759172
C1=CN=CC=C1CC2=CN=CN2.Br.Br
Specifications
Immethridine dihydrobromide
699020-93-4
immethridine dihydrobromide, 4-1h-imidazol-4-ylmethyl pyridine dihydrobromide, 4-1h-imidazol-5-ylmethyl pyridine;dihydrobromide, 4-1h-imidazol-5-yl methyl pyridine-hydrogen bromide 1/2
C1=CN=CC=C1CC2=CN=CN2.Br.Br
321.016
321.01
10mg
C9H11Br2N3
CYNKWHIKNDIVDR-UHFFFAOYSA-N
4-(1H-imidazol-5-ylmethyl)pyridine;dihydrobromide
16759172
>99%
Videos