Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.80000kg
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
- Mesh bottom
- One handle on each end
- Functional and modern design
- Available for a variety of capacities
Specifications
Basket | |
Multi-Frequency Ultrasonic Cleaner, TI-H-160 135 L | |
Stainless Steel |