Learn More
Tocris Bioscience™ Immepip dihydrobromide
Standard H3 agonist. Also H4 agonist
Brand:  Tocris Bioscience™ 0932/50
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13448173
  • £630.63 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Potent histamine H3 receptor agonist. Also binds to H4 receptors (Ki values are 0.4 and 9 nM at human recombinant H3 and H4 receptors respectively). Equipotent to or slightly more active than (R)-α-methylhistamine at H3 receptors.
Specifications
Specifications
4-(1H-Imidazol-4-ylmethyl)piperidine dihydrobromide
C9H15N3.2HBr
50mg
164391-47-3
Observed MW: 327.06
Soluble to 100mM in water