Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Ovalbumin
Prepare hapten-carrier conjugates to generate antibodies against cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive ovalbumin protein.
Brand:  Thermo Scientific™ 77125
UNSPSC : 41106500
Code : PN
Additional Details : CAS Number : 6769-80-8 Weight : 0.80000kg
 View more versions of this product
Product Code. 11894061
  • £306.00 / 10mg
Estimated Shipment
17-07-2017
Add to basket
Specifications
Description
Purified, carrier-grade ovalbumin has been modified with SMCC crosslinker to attach maleimide groups capable of forming covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other molecules. Immunogens are easily prepared from peptide antigens that have been engineered with a terminal cysteine residue as a precise conjugation point. Several antigen molecules can be attached per carrier protein molecule without blocking intended epitopes by crosslinking to primary amines or carboxylates within the peptide sequence.
Highlights:
  • Maleimide-Activated Ovalbumin, supplied lyophilized in PBS (pH 7.2) with stabilizer
  • Activated with 5 to 15 moles of maleimide per mole of ovalbumin
  • Ideal for conjugating cysteine-containing peptide antigens to make immunogen or screening target
  • Conjugation reaction complete in 2 hours; results in stable, covalent thioether bonds
  • Single polypeptide protein with molecular mass of 45,000 daltons
  • Contains 14 aspartic acid and 33 glutamic acid residues whose carboxyl groups account for the net negative charge (pI 4.63) of OVA
  • Not as immunogenic as keyhole limpet hemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent
  • Useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
Description & Specifications
Specifications
Activated protein lyophilized in PBS (pH 7.2) with stabilizer
Lyophilized
OVA, ovalbumin
10mg
5 conjugation reactions, each with up to 2mg sulfhydryl-containing peptide
Imject Maleimide-Activated Ovalbumin
Maleimide