missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, 97%, Thermo Scientific Chemicals

Catalog Number 10563141
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
256-96-2
C14H11N
193.25
MFCD00005071
LCGTWRLJTMHIQZ-UHFFFAOYSA-N
iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
9212
CHEBI:47802
11H-benzo[b][1]benzazepine
C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2
This item is not returnable. View return policy
256-96-2
C14H11N
193.25
MFCD00005071
LCGTWRLJTMHIQZ-UHFFFAOYSA-N
iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
9212
CHEBI:47802
11H-benzo[b][1]benzazepine
C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 197°C to 201°C
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 193.25
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Iminostilbene, 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'