missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iminostilbene, 97%, Thermo Scientific Chemicals
27.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.
Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.
Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 197°C to 199°C |
Quantity | 25 g |
Beilstein | 1343358 |
Solubility Information | Soluble in ethyl acetate (25mg/ml),water (partly),dioxane,chloroform,and DMSO. |
Formula Weight | 193.25 |
Percent Purity | 97% |
Chemical Name or Material | Iminostilbene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'