missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iminodibenzyl, 97%, Thermo Scientific Chemicals
14.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 105°C to 108°C |
Color | Beige to Yellow |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 25 g |
Formula Weight | 195.26 |
Percent Purity | 97% |
Physical Form | Crystalline Powder |
Show More |
Safety and Handling
Product Identifier
- Iminodibenzyl
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'