missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Iminodiacetic acid, 98+%
11.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 243°C |
Color | White or Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Plastic bottle |
Linear Formula | HN(CH2CO2H)2 |
Quantity | 25 g |
Beilstein | 04, 365 |
Merck Index | 15, 4959 |
Solubility Information | Solubility in water: 2.43g/100 mL water (5°C). Other solubilities: 2.9g/100 mL water (20°C),6.3g/100 mL water (40°C),12.8g/100 mL water (60°C),23.0g/100 mL water (80°C),32.5g/100 mL water (100°C) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'