missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Imide-extended bismaleimide, BMI-5000

£70.20 - £282.00

Chemical Identifiers

CAS 921213-77-6
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492687
SDS View Documents
Alfa Aesar™
046460.14
25g
£70.20
25g
View alternative products
Save Up to 
»
Estimated Shipment: 25-10-2021
Add to basket
 
15402697
SDS View Documents
Alfa Aesar™
046460.22
100g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

921213-77-6

Specifications

Imide-extended bismaleimide
921213-77-6
Yellow
BMI-5000
Powder
25g
SDS
Product Certifications