missing translation for 'onlineSavingsMsg'
Learn More

Imidazolidinyl urea, Thermo Scientific Chemicals

Catalog Number p-7040846
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
39236-46-9
C11H16N8O8
388.297
MFCD00038890
ZCTXEAQXZGPWFG-UHFFFAOYSA-N
imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
38258
CHEBI:51805
1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO
This item is not returnable. View return policy
39236-46-9
C11H16N8O8
388.297
MFCD00038890
ZCTXEAQXZGPWFG-UHFFFAOYSA-N
imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
38258
CHEBI:51805
1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 141°C to 143°C
Quantity 25 g
Formula Weight 388.3
Chemical Name or Material Imidazolidinyl urea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'