missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, sodium derivative, ACROS Organics™

Brand:  Acros Organics 188980050

Code : 29

Additional Details : CAS Number : 5587-42-8 Weight : 0.05500kg Transport : UN number : 2923 Chem class : 8 Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10262842

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Imidazole, sodium derivative
5587-42-8
C3H3N2Na
MFCD00064314
Sodium derivative
≥97.0%
90.06

Specifications

Imidazole, sodium derivative
Sodium derivative
Authentic
Glass bottle
≥97.0%
90.06
23, II, 35
5g
T1130
5587-42-8
3% max.
Free Flowing Crystalline Powder
C3H3N2Na
MFCD00064314
Light yellow to orange
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Imidazole, sodium derivative

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS