missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, sodium derivative, ACROS Organics™

Brand:  Acros Organics 188980250

Code : 29

Additional Details : CAS Number : 5587-42-8 Weight : 0.07500kg Transport : UN number : 2923 Chem class : 1b Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10028841

  • £100.30 / 25g


Estimated Shipment
14-02-2018
Add to basket


Specifications

Chemical Identifiers

Imidazole, sodium derivative
5587-42-8
C3H3N2Na
MFCD00064314
Sodium derivative
≥97.0%
90.06

Specifications

Imidazole, sodium derivative
Sodium derivative
Authentic
Glass bottle
≥97.0%
90.06
23, II, 35
25g
T1130
5587-42-8
3% max.
Free Flowing Crystalline Powder
C3H3N2Na
MFCD00064314
Light yellow to orange
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Imidazole, sodium derivative

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS