missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 162.15 |
Percent Purity | 97% |
Chemical Name or Material | Imidazo[1, 2-a]pyridine-6-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'