missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Packaging | Amber Glass Bottle |
Linear Formula | Yes |
Quantity | 250 mg |
Formula Weight | 217.05 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Imidazo[1, 2-a]pyridine-6-carbonyl chloride hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'